Specifications
- Description:Aquatest 2010 Karl-Fischer Coulometric Moisture Titrator
- Cat. No.:55577-206
- Supplier no.:0092010
Specifications
About this item
The Photovolt Aquatest 2010 moisture analyzer provides rapid, accurate convenient low level moisture analysis from low ppm range to pure water with maximum isolation of solutions from atmospheric moisture
With a range of 1 ppm to 1,000,000, typical Aquatest 2010 applications include low level moisture applications for liquids and dispersed solids, pharmaceutical samples, lubricating oils, chemicals and solvents, resins, lyophilized samples and plastics.
The Department of Defense Naval Air Systems Command (NAVAIR) recommends the Photovolt Aquatest 2010 as the physical test and laboratory operating procedure for their Joint Oil Analysis Program for the navy, army, air force and coast guard.
Available with a diaphragm type or diaphragmless generator cartridge, the Aquatest 2010 is a full-featured, user-friendly operating system with multiple RS-232C ports for system expansion for simultaneous interface with both a balance and a computer.