About this item
For the isolation of microorganisms from surfaces sanitized with quaternary ammonium compounds.
- Isolate microorganisms from sanitized surfaces
- With lechitin and polysorbate 80
Tryptic soy agar with lecithin and polysorbate 80 is used for the isolation of microorganisms from surfaces sanitized with quaternary ammonium compounds.