Anti-SRP9 Mouse Monoclonal Antibody [clone: 1H7G10]
Catalog # 10095-178
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Signal Recognition Particle 9kDa
- Antigen Symbol:SRP9
- Clonality:Monoclonal
- Clone:1H7G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein A Purification
- Cat. No.:10095-178
- Supplier no.:66068-1-IG
Specifications
About this item
Signal recognition particle 9(SRP9) is component of the signal recognition particle (SRP), which is a ribonucleoprotein complex that mediates the targeting of proteins to the rough endoplasmic reticulum (ER). SRP9 form a heterodimer with SRP14, which involves in arrest of the elongation of the nescent chains during targeting to ensure efficient translocation of the preprotein.
Western Blot: Hela Cells, 1:200-1:2000
Type: Primary
Antigen: SRP9
Clonality: Monoclonal
Clone: 1H7G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human