Anti-CD11c/Integrin alpha X Mouse Monoclonal Antibody [clone: 2F1C10]
Catalog # 10089-012
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Integrin Subunit Alpha X
- Antigen Symbol:ITGAX
- Clonality:Monoclonal
- Clone:2F1C10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Size:150 µL
- Antigen Synonyms:CD11C|SLEB6
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Protein A Purification
- Cat. No.:10089-012
- Supplier no.:60258-1-IG
Specifications
About this item
Integrins are cell adhesion receptors that are heterodimers composed of non-covalently associated α and β subunits. CD11c/Integrin alpha X is a type I transmembrane protein present on a variety of cells, including monocytes/macrophages, granulocytes, NK cells and dendritic cells. Integrin alpha X/beta 2 acts a receptor for fibrinogen and is important in monocyte adhesion and chemotaxis.
IHC: Human tonsillitis Tissue, 1:20-1:200
Type: Primary
Antigen: CD11c/Integrin alpha X
Clonality: Monoclonal
Clone: 2F1C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse