Anti-Connexin-26 Rabbit Polyclonal Antibody
Catalog # 10082-036
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Connexin 26
- Antigen Symbol:Cx26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10082-036
- Supplier no.:51037-2-AP
Specifications
About this item
Western Blot: Human Brain Tissue, 1:200-1:1000; IHC: Human Colon cancer Tissue, 1:20-1:200
Type: Primary
Antigen: Connexin-26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat