Anti-ATXN3L Rabbit Polyclonal Antibody
Catalog # 10083-406
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Ataxin 3-like
- Antigen Symbol:ATXN3L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:ATXN3L
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10083-406
- Supplier no.:25099-1-AP
Specifications
About this item
ATXN3L also named as ataxin 3 like or MJDL is a 355 amino acid protein, which contains one Josephin domain and two UIM repeats. ATXN3L localizes in nucleus and may cleave poly ubiquitin chain.
Western Blot: Human Testis Tissue, 1:200-1:2000
Type: Primary
Antigen: ATXN3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse