To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:C15orf53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10083-776
- Supplier no.:24553-1-AP
Specifications
About this item
C15orf53 has been identified on chromosome 15q14. It is probably neither causative for the etiology of BD (Bipolar disorder) nor for SCZD10 (periodic catatonia). Gene expression analysis revealed that C15orf53 was expressed in a subpopulation of leukocytes, but not in human post-mortem limbic brain tissue. In our detection this antibody always detect ~30 kDa band even the amino acids is 20 kDa.
IHC: Human tonsillitis Tissue, 1:20-1:200; Western Blot:A431 Cells, 1:500-1:5000
Type: Primary
Antigen: C15orf53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human