Anti-PRPF39 Rabbit Polyclonal Antibody
Catalog # 10092-828
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Pre-mRNA Processing Factor 39
- Antigen Symbol:PRPF39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10092-828
- Supplier no.:24480-1-AP
Specifications
About this item
PRPF39 also termed as Pre mRNA processing factor 39 or PRP39 homolog is a 669 amino acid protein, which contains 7 HAT repeats and belongs to the PRP39 family. PRPF39 localizes in nucleus and may play a role in mRNA splicing.
Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200
Type: Primary
Antigen: PRPF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human