Order Entry
United States
ContactUsLinkComponent
Anti-MFSD8 Rabbit Polyclonal Antibody
Anti-MFSD8 Rabbit Polyclonal Antibody
Catalog # 10089-452
Supplier:  Proteintech
undefined
Anti-MFSD8 Rabbit Polyclonal Antibody
Catalog # 10089-452
Supplier:  Proteintech
Supplier Number:  24298-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Major Facilitator Superfamily Domain Containing 8
  • Antigen Symbol:
    MFSD8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10089-452
  • Supplier no.:
    24298-1-AP

Specifications

About this item

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: MFSD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse