Anti-IGHMBP2 Rabbit Polyclonal Antibody
10088-670
- Antibody Type:Primary
- Antigen Name:Immunoglobulin Helicase Mu-binding Protein 2
- Antigen Symbol:IGHMBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Immunoglobulin S mu binding protein 2|Immunoglobulin mu binding protein 2|CATF1|IGHMBP 2|Immunoglobulin mu-binding protein 2|RIPE3 b1|AEP|DNA-binding protein SMUBP-2|Antreeze enhancer binding protein|SMBP2_HUMAN|GF-1|HMN 6|Cardiac transcription factor1|ATP-dependent helicase IGHMBP2|Glial factor 1|SMARD1|RIPE3b 1|SMUBP 2.|Cardiac transcription factor 1|SMARD 1|Immunoglobulin mu binding protein2|Ighmbp2|CATF 1|HCSA|HMN6|RIPE3b1|Immunoglobulin S mu binding protein2
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. No.:10088-670
- Supplier no.:23945-1-AP
IGHMBP2 also named as immunoglobulin mu binding protein 2 is a 993 amino-acid protein, which localizes in nuclear and cytoplasm. The molecular weight of IGHMBP2 is 109 kDa. IGHMBP2 belongs to the DNA2/NAM7 helicase family. IGHMBP2 also acts as a transcriptional regulator and is necessary for transcriptional activation of the flounder liver-type antifreeze protein gene. IGHMBP2 exists as a homooligomer and is part of the cytosolic ribonucleoprotein complex.
Western Blot: HL-60 Cells, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200
Type: Primary
Antigen: IGHMBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human