Anti-CEP55 Rabbit Polyclonal Antibody
Catalog # 10084-654
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Centrosomal protein 55kDa
- Antigen Symbol:CEP55
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:URCC6|CT111|C10orf3
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. No.:10084-654
- Supplier no.:23891-1-AP
Specifications
About this item
CEP55, also named as C10orf3 and URCC6, is a 55 kDa protein which located in the centrosome in interphase cells and recruited to the midbody during cytokinesis, is essential for completion of cell abscission. It plays a role in mitotic exit and cytokinesis.
Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200
Type: Primary
Antigen: CEP55
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: