Order Entry
United States
ContactUsLinkComponent
Anti-FOXQ1 Rabbit Polyclonal Antibody
Anti-FOXQ1 Rabbit Polyclonal Antibody
Catalog # 10087-136
Supplier:  Proteintech
undefined
Anti-FOXQ1 Rabbit Polyclonal Antibody
Catalog # 10087-136
Supplier:  Proteintech
Supplier Number:  23718-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Forkhead Box Q1
  • Antigen Symbol:
    FOXQ1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    HFH1|FKHRL1
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen Affinity Purified
  • Cat. No.:
    10087-136
  • Supplier no.:
    23718-1-AP

Specifications

About this item

Forkhead box Q1 (FOXQ1, also known as HFH1), a member of the FOX gene family, contains the core DNA binding domain, whereas the flanking wings of FOXQ1 contribute to its sequence specificity. FOXQ1 is known to repress the promoter activity of smooth muscle–specific genes, such as telokin and SM22α. FOXQ1 expression is regulated by Hoxa1 in embryonic stem cells. As a transcription factor, FOXQ1 represses the promoter activity of smooth muscle–specific genes, such as telokin and SM22α, in A10 vascular muscle cells .

Western Blot: Mouse Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: FOXQ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat