- Antibody Type:Primary
- Antigen Name:T-box 18
- Antigen Symbol:TBX18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:T-box protein 14|T-box transcription factor TBX15|T-box protein 15|T-box transcription factor TBX18|TBX15|TBX14|T-box protein 18|T-box transcription factor TBX14
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. No.:10095-660
- Supplier no.:23237-1-AP
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator. The Tbx genes constitute a family of transcriptional regulatory genes that are implicated in a variety of developmental processes ranging from the formation of germ layers to the organizational patterning of the central nervous system.This antibody specifically reacts with the 65kd TBX18 protein.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200
Type: Primary
Antigen: TBX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat