Specifications
- Antibody Type:Primary
- Antigen Name:Epithelial Splicing Regulatory Protein B
- Antigen Symbol:ESRRB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:NR3B2|DFNB35|ESRL2|ERR2|ERRb
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10086-666
- Supplier no.:22644-1-AP
Specifications
About this item
Estrogen related receptor beta (ESRRB), a NR3 Steroid Receptor, has been shown to affect early placental development. Also it can act as a repressor of transcriptional activity mediated by the glucocorticoid receptor (GR). It binds as a monomer to the extended half-site TNAAGTGCA and as a homodimer to the estrogen response element (ERE), palindromic thyroid hormone response element (TRE(pal)), and SF-1 response element, but not to the glucocorticoid response element (GRE). Estrogen related receptor beta mutations lead to abnormal development of the chorion and defective diploid trophoblast proliferation, and are lethal during midgestation. Expression has been documented in mice in the extra embryonic ectoderm that gives rise to the chorion. ESTs have been isolated from normal human eye, lung, and testis libraries. The ligand for the receptor is PPARgamma coactivator 1 (PGC-1) beta.
Western Blot:PC-13 Cells, 1:500-1:5000
Type: Primary
Antigen: ESRRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat