Anti-Syntenin 1 Rabbit Polyclonal Antibody
Catalog # 10095-542
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Syntenin 1
- Antigen Symbol:SDCBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10095-542
- Supplier no.:22399-1-AP
Specifications
About this item
Western Blot: HepG2 Cells, 1:500-1:5000; IP: HeLa Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200; FC: HepG2 Cells, N/A
Type: Primary
Antigen: Syntenin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human