Anti-DCP1A Rabbit Polyclonal Antibody
Catalog # 10085-704
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Decapping mRNA 1A
- Antigen Symbol:DCP1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Smad4-interacting transcriptional co-activator|decapping enzyme |DCP1A|Nbla00360|HSA275986|putative protein product of Nbla00360|SMIF|decapping enzyme hDcp1a|transcription factor SMIF|SMAD4IP1
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. No.:10085-704
- Supplier no.:22373-1-AP
Specifications
About this item
DCP1 decapping enzyme homolog A (S. cerevisiae), known as mRNA-decapping enzyme 1A (DCP1A),is necessary for the degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA molecules, yielding a 5'-phosphorylated mRNA fragment and 7m-GDP. Contributes to the transactivation of target genes after stimulation by TGFB1. This antibody specifically react with the 70kd DCP1A protein.
Western Blot: Mouse lung Tissue, 1:500-1:5000; IHC: Mouse lung Tissue, 1:20-1:200
Type: Primary
Antigen: DCP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse