Specifications
- Antibody Type:Primary
- Antigen Name:Centrosomal Protein 97kDa
- Antigen Symbol:CEP97
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:CEP 97|RGD1307400.|Leucine rich repeat and IQ domain containing protein 2|LRRIQ2|Centrosomal protein of 97 kDa|Centrosomal protein 97kDa|Leucine rich repeats and IQ mot containing 2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10084-664
- Supplier no.:22050-1-AP
Specifications
About this item
CEP97, also named as LRRIQ2, contains one IQ domain, eight LRR (leucine-rich) repeats and one LRRCT domain. It collaborates with CEP110, being involved in the suppression of a cilia assembly program. CEP97 is required for correct spindle formation and has a role in cytokinesis. It is required for recruitment of CEP110 to the centrosome. This protein has a calculated MW ~97 kDa, although the protein migrates at a molecular weight of approximate 140 kDa. The protein is highly acidic, with a pI ~4.7, providing a potential explanation for its reduced mobility in SDS-PAGE. . This is a rabbit polyclonal antibody raised against the C terminus of human CEP97.
Western Blot: HL-60 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: MDCK Cells, 1:20-1:200
Type: Primary
Antigen: CEP97
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: