Anti-PVRL4 Rabbit Polyclonal Antibody
Catalog # 10093-106
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Nectin Cell Adhesion Molecule 4
- Antigen Symbol:NECTIN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:nectin-4|EDSS1|PRR4|LNIR
- Storage Buffer:PBS with 0.0.2% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity Purified
- Cat. No.:10093-106
- Supplier no.:21903-1-AP
Specifications
About this item
Nectin-4, or PVRL4, belongs to the family of cell adhesion molecules that regulate the formation of adherence functions in epithelial cells through the AF-6 afadin scaffold molecule. Nectin-4 is reported as a histological and serological marker for breast carcinoma.
Western Blot: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: PVRL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse