Order Entry
United States
ContactUsLinkComponent
Anti-ZO1 Rabbit Polyclonal Antibody
Anti-ZO1 Rabbit Polyclonal Antibody
Catalog # 10097-394
Supplier:  Proteintech
Anti-ZO1 Rabbit Polyclonal Antibody
Catalog # 10097-394
Supplier:  Proteintech
Supplier Number:  21773-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Zona occludens 1
  • Antigen Symbol:
    ZO1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10097-394
  • Supplier no.:
    21773-1-AP

Specifications

About this item

Tight junction (or zonula occludens) form the continuous intercellular barrier between epithelial and endothelial cells, which is required to separate tissue spaces and regulate selective movement of solutes across the epithelium and endothelium . ZO-1 (also known as TJP1) is a peripheral membrane phosphoprotein located on the cytoplasmic face and is expressed in tight junctions of both epithelial and endothelial cells . It binds the transmembrane proteins occludin and the claudins linking them to cytoskeletal actin . ZO-1 belongs to a family of multidomain proteins known as the membrane-associated guanylate kinase homologs (MAGUKs). It is a pivotal tight junction protein and may be involved in signalling mechanisms regulating cell proliferation and differentiation .

Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A; IF: HUVEC Cells, 1:20-1:200

Type: Primary
Antigen: ZO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human