Anti-ZEB2 Rabbit Polyclonal Antibody
10094-396
- Antibody Type:Primary
- Antigen Name:Zinc Finger E-box Binding Homeobox 2
- Antigen Symbol:ZEB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:HSPC082|Smad-interacting protein 1|SIP1|ZFX1B|KIAA0569|Zinc finger E-box-binding homeobox 2|SMADIP1|ZFHX1B|SIP-1|HRIHFB2411|ZEB2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10094-396
- Supplier no.:21672-1-AP
Western Blot: HepG2 Cells, 1:500-1:5000
Type: Primary
Antigen: ZEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat