Order Entry
ContactUsLinkComponent
Anti-ENTPD8 Rabbit Polyclonal Antibody
Anti-ENTPD8 Rabbit Polyclonal Antibody
Catalog # 10081-296
Supplier:  Proteintech
undefined
Anti-ENTPD8 Rabbit Polyclonal Antibody
Catalog # 10081-296
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ectonucleoside triphosphate diphosphohydrolase 8
  • Antigen Symbol:
    ENTPD8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    E-NTPDase|UNQ2492|GLSR2492|NTPDase-8
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10081-296
  • Supplier no.:
    21010-1-AP

Specifications

About this item

ENTPD8(Ectonucleoside triphosphate diphosphohydrolase 8) is also named as E-NTPDase 8, NTPDase8 and belongs to the GDA1/CD39 NTPase family. NTPDase8 is the canalicular ecto-ATPase/ATPDase and is responsible for the main hepatic NTP-Dase activity. The estimated molecular mass of expressed human E-NTPDase 8 is 70-85 kDa which is N-glycosylated and after deglycosylation,it has a nearly identical molecular mass, 54 kDa. It has 2 isoforms produced by alternative splicing with the molecular weight of 54 kDa and 49 kDa. This antibody is specific to ENTPD8.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ENTPD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Frequently Bought Together