Anti-ALG6 Rabbit Polyclonal Antibody
Catalog # 10082-790
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:ALG8, alpha 1,3-glucosyltransferase
- Antigen Symbol:ALG6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:ALG6
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-790
- Supplier no.:20753-1-AP
Specifications
About this item
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: ALG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat