Anti-AGPAT9 Rabbit Polyclonal Antibody
Catalog # 10082-610
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:1-Acylglycerol-3-Phosphate O-Acyltransferase 9
- Antigen Symbol:AGPAT9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-610
- Supplier no.:20603-1-AP
Specifications
About this item
Western Blot:rat Kidney Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: AGPAT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat