Order Entry
ContactUsLinkComponent
Anti-IGF1R Rabbit Polyclonal Antibody
Anti-IGF1R Rabbit Polyclonal Antibody
Catalog # 10088-650
Supplier:  Proteintech
undefined
Anti-IGF1R Rabbit Polyclonal Antibody
Catalog # 10088-650
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Insulin-like Growth Factor 1 Receptor
  • Antigen Symbol:
    IGF1R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    MGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10088-650
  • Supplier no.:
    20254-1-AP

Specifications

About this item

IGF1R, also named as CD221, belongs to the protein kinase superfamily, Tyr protein kinase family and Insulin receptor subfamily. IGF1R binds insulin-like growth factor 1 (IGF1) with a high affinity and IGF2 with a lower affinity. It has a tyrosine-protein kinase activity, which is necessary for the activation of the IGF1-stimulated downstream signaling cascade. When present in a hybrid receptor with INSR, IGF1R binds IGF1. The hybrid receptors composed of IGF1R and INSR isoform Long are activated with a high affinity by IGF1, with low affinity by IGF2 and not significantly activated by insulin, and that hybrid receptors composed of IGF1R and INSR isoform Short are activated by IGF1, IGF2 and insulin. In contrast, the hybrid receptors composed of IGF1R and INSR isoform Long and hybrid receptors composed of IGF1R and INSR isoform Short have similar binding characteristics, both bind IGF1 and have a low affinity for insulin. Defects in IGF1R may be a cause in some cases of resistance to insulin-like growth factor 1 (IGF1 resistance). This antibody raised against a synthesized polypeptide (CGVLVLRASFDERQPYAH) corresponding to 1332-1348aa of human IGF1R protein recognizes beta chain of IGF1R.

Western Blot: HeLa Cells, 1:500-1:5000; IP: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Frequently Bought Together