To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Thymosin Beta 4, X-linked
- Antigen Symbol:TMSB4X
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:TB4X|TMSB4|FX|PTMB4
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-108
- Supplier no.:19850-1-AP
Specifications
About this item
Thymosin beta 4 (Tβ4), encoded by TMSB4X gene, is an actin sequestering protein which plays an important role in the organization of the cytoskeleton. Numerous studies have demonstrated the effects of Tβ4 on cell migration, proliferation, apoptosis and inflammation after exogenous treatment . Recently, novel findings provided compelling evidence that Tβ4 played a key role in facilitating tumor metastasis and angiogenesis. It has been found that Tβ4 expressed increasingly in a number of metastatic tumors, thus providing a potential target of opportunity for cancer management, especially for cancer metastasis therapy .
Western Blot: Human Skeletal Muscle Tissue, 1:1000-1:10000
Type: Primary
Antigen: TMSB4X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat