Order Entry
United States
ContactUsLinkComponent
Anti-RtcB Rabbit Polyclonal Antibody
Anti-RtcB Rabbit Polyclonal Antibody
Catalog # 10083-840
Supplier:  Proteintech
Anti-RtcB Rabbit Polyclonal Antibody
Catalog # 10083-840
Supplier:  Proteintech
Supplier Number:  19809-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    RtcB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10083-840
  • Supplier no.:
    19809-1-AP

Specifications

About this item

RTCB (also known as HSPC117, C22orf28, FAAP and D10Wsu52e) is an essential subunit of a tRNA ligase complex that is involved in RNA repair and stress-induced splicing. As a multifunctional protein, RTCB is also a cell adhesion protein and is important in embryo and placenta development. Recently RTCB has been identified as UPR RNA ligase catalyzing unconventional XBP1 mRNA splicing. This antibody specifically recognizes endogenous RTCB protein. (25087875)

Western Blot: HeLa Cells, 1:1000-1:10000; IF: HeLa Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: RtcB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse