Anti-KALRN Rabbit Polyclonal Antibody
Catalog # 10089-102
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Kalirin RhoGEF kinase
- Antigen Symbol:KALRN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Huntingtin-associated protein-interacting protein|RhoGEF kinase|Huntingtin associated protein interacting protein duo |Protein Duo|Kalirin|Duo|HAPIP|TRAD.|Serine/threonine kinase with Dbl and pleckstrin homology domains |KALRN_HUMAN |Kalirin isoform 2|KALRN |Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain|FLJ16443
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10089-102
- Supplier no.:19740-1-AP
Specifications
About this item
KALRN, also named as DUET, DUO, HAPIP and TRAD, belongs to the protein kinase superfamily and CAMK Ser/Thr protein kinase family. KALRN promotes the exchange of GDP by GTP. KALRN activates specific Rho GTPase family members, thereby inducing various signaling mechanisms that regulate neuronal shape, growth, and plasticity, through their effects on the actin cytoskeleton. KALRN induces lamellipodia independent of its GEF activity. The antibody can recognize isoform1 and isoform2 of KALRN.
Western Blot: Human Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse