Anti-SPTBN1 Rabbit Polyclonal Antibody
Catalog # 10095-130
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Spectrin Beta Non-erythrocytic 1
- Antigen Symbol:SPTBN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:betaSpII|HEL102|ELF|SPTB2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10095-130
- Supplier no.:19722-1-AP
Specifications
About this item
SPTBN1 is the non-erythrocytic form of β-spectrin or β-fodrin. Spectrins are members of the superfamily of F-actin cross linking proteins that are important as scaffolding proteins for protein sorting, cell adhesion, and migration. Reduced expression of SPTBN1 has been found to correlate with shorter survival of patients with hepatocellular cancer, pancreatic cancer and other malignancies of the gastrointestinal tract, suggesting tumor suppression ability of this protein.
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200; IP: HEK-293 Cells, 1:200-1:2000
Type: Primary
Antigen: SPTBN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse