Anti-Histone H1.2 Rabbit Polyclonal Antibody
Catalog # 10088-270
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Histone H1.2
- Antigen Symbol:Histone H1.2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10088-270
- Supplier no.:19649-1-AP
Specifications
About this item
Histone H1 regulates chromatin competency through its high affinity binding to linker DNA as a structural component. It acts also as a regulator of individual gene transcription through chromatin remodeling, nucleosome spacing and DNA methylation.
Western Blot:L02 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: Histone H1.2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat