Anti-NY-ESO-1 Rabbit Polyclonal Antibody
Catalog # 10091-512
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:NY-ESO-1
- Antigen Symbol:NY-ESO-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10091-512
- Supplier no.:19521-1-AP
Specifications
About this item
CTAG1A, also named as NY-ESO-1, CTAG, CTAG1, ESO1, LAGE2, LAGE2A, CTAG1B, LAGE2B and CT6.1, belongs to the CTAG family.
Western Blot: Human Testis Tissue, 1:200-1:1000
Type: Primary
Antigen: NY-ESO-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human