Order Entry
United States
ContactUsLinkComponent
Anti-DNA2 Rabbit Polyclonal Antibody
Anti-DNA2 Rabbit Polyclonal Antibody
Catalog # 10085-582
Supplier:  Proteintech
undefined
Anti-DNA2 Rabbit Polyclonal Antibody
Catalog # 10085-582
Supplier:  Proteintech
Supplier Number:  18727-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    DNA Replication Helicase/nuclease 2
  • Antigen Symbol:
    DNA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    150 µL
  • Antigen Synonyms:
    DNA2L|hDNA2
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10085-582
  • Supplier no.:
    18727-1-AP

Specifications

About this item

DNA2(DNA replication ATP-dependent helicase/nuclease DNA2) is also named as DNA2L, KIAA0083 and belongs to the DNA2/NAM7 helicase family. This protein includes 2 domains: DNA replication nuclease and DNA replication ATP-dependent helicase. It is a conserved helicase/nuclease involved in the maintenance of mitochondrial and nuclear DNA stability and it can interact with DNA polymerase gamma and stimulating its activity. It can also process intermediate 5' flap structures occuring in DNA replication long-patch base excision repair in mitochondria. It has 4 isoforms produced by alternative splicing. This antibody is specific to DNA2.

IHC: Human cervical cancer, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DNA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human