Order Entry
United States
ContactUsLinkComponent
Anti-RPS9 Rabbit Polyclonal Antibody
Anti-RPS9 Rabbit Polyclonal Antibody
  10093-988
undefined
Anti-RPS9 Rabbit Polyclonal Antibody
  10093-988
 :  18215-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Ribosomal Protein 9
  • Antigen Symbol:
    RPS9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    40S ribosomal protein S9|RPS9
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10093-988
  • Supplier no.:
    18215-1-AP

 

 

Western Blot: Mouse Uterus Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: RPS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse