Order Entry
United States
ContactUsLinkComponent
Anti-PRPH2 Rabbit Polyclonal Antibody
Anti-PRPH2 Rabbit Polyclonal Antibody
Catalog # 10092-838
Supplier:  Proteintech
undefined
Anti-PRPH2 Rabbit Polyclonal Antibody
Catalog # 10092-838
Supplier:  Proteintech
Supplier Number:  18109-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Peripherin 2
  • Antigen Symbol:
    PRPH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    PRPH2_HUMAN|Peripherin-2|Retinal degeneration slow protein|Tspan-22.|Tetraspanin-22|PRPH2
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10092-838
  • Supplier no.:
    18109-1-AP

Specifications

About this item

Peripherin-2 (PRPH2), also known as retinal degeneration slow protein (RDS), is a photoreceptor-specific tetraspanin protein implicated in outer segment disk morphogenesis. It may function as an adhesion molecule involved in stabilization and compaction of outer segment disks or in the maintenance of the curvature of the rim. Mutations in peripherin-2 are responsible for various retinal degenerative diseases including autosomal dominant retinitis pigmentosa (ADRP).

Western Blot: Mouse Eye Tissue, 1:200-1:2000; IF: Mouse Retina Tissue, 1:20-1:200

Type: Primary
Antigen: PRPH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat