Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 21
- Antigen Symbol:USP21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-852
- Supplier no.:17856-1-AP
Specifications
About this item
USP21(Ubiquitin carboxyl-terminal hydrolase 21) is also named as USP23.The USPs belong to a subclass of the protein-deubiquitinating enzyme (DUB) superfamily that are categorized into five subclasses based on their ubiquitin-protease domains in the human genome and have been shown to be involved in a broad range of biological activities. USP21 is expressed at different levels in the tissues examined. The highest level of USP21 expression is found in the heart, pancreas, and skeletal muscle. USP21 messages could also be detected in the brain, placenta, liver, and kidney but are very low in the Lung through the northern blot.
Western Blot:recombinant ptotein, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200
Type: Primary
Antigen: USP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human