To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Acyl-CoA Dehydrogenase Long Chain
- Antigen Symbol:ACADL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:ACAD4|LCAD
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10082-392
- Supplier no.:17526-1-AP
ACADL(Long-chain specific acyl-CoA dehydrogenase, mitochondrial) is also named as LCAD and belongs to the acyl-CoA dehydrogenase family.It catalyzes the first reaction of the mitochondrial β-oxidation of fatty acids and is synthesized in the cytosol as a precursor that is larger than its mature from. The size difference between the precursor and mature rat LCAD has been estimated to be 3 kDa on the basis of SDS-PAGE.Defects in ACADL are a cause of acyl-CoA dehydrogenase very long-chain deficiency (ACADVLD).This antibody is specific to ACADL.
Western Blot: Mouse Kidney Tissue, 1:200-1:2000; IHC: Human Colon Tissue, 1:20-1:200
Type: Primary
Antigen: ACADL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat