- Antibody Type:Primary
- Antigen Symbol:EIF4A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10086-438
- Supplier no.:17504-1-AP
EIF4A3 is a component of the exon junction complex (EJC), which assembles near exon-exon junctions of mRNAs as a result of splicing. EJC proteins involves in postsplicing events, including mRNA export, cytoplasmic localization, and nonsense-mediated decay. Its RNA-dependent ATPase and RNA-helicase activities are induced by CASC3, but abolished in presence of the MAGOH/RBM8A heterodimer, thereby trapping the ATP-bound EJC core onto spliced mRNA in a stable conformation. Besides, it involved in translational enhancement of spliced mRNAs after formation of the 80S ribosome complex and binds spliced mRNA in sequence-independent manner, 20-24 nucleotides upstream of mRNA exon-exon junctions
Western Blot:A549 Cells, 1:1000-1:10000; IHC: Human Kidney Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100; IP: Mouse Heart Tissue, 1:500-1:5000
Type: Primary
Antigen: EIF4A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat