- Antibody Type:Primary
- Antigen Name:Fatty Acid Binding Protein 7, brain
- Antigen Symbol:FABP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:BLBP|LTR2-FABP7|FABPB|B-FABP|MRG
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10086-406
- Supplier no.:17456-1-AP
FABP7 (Fatty Acid Binding Protein-7), also known as B-FABP (Brain Fatty Acid-Binding Protein), is a member of FABP family which are intracellular proteins involved in lipid metabolism. Normally FABP7 is mainly expressed in brain and other neural tissues and is important in early nervous system development. In addition, FABP7 is considered a marker for some brain tumors and may have a role in tumorigenesis in the peripheral nervous system and in several extra nervous system neoplasms such as breast cancer, renal cancer, and melanoma. This antibody was raised against the full-length human FABP7 and may cross-react with other FABPs.
Western Blot: Human Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: FABP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat