Order Entry
United States
Orders LinkContactUsLinkComponent
Anti-CD33 Rabbit Polyclonal Antibody

Anti-CD33 Rabbit Polyclonal Antibody

150 µL
Catalog # 10084-332
Anti-CD33 Rabbit Polyclonal Antibody
Catalog # 10084-332
Supplier:  
Anti-CD33 Rabbit Polyclonal Antibody
Anti-CD33 Rabbit Polyclonal Antibody

150 µL

Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CD33, aslo referred to as Siglec-3, is a 67-kDa glycosylated transmembrane protein of sialic acid-binding immunoglobulin-like lactic (siglec) family. CD33 expression is restrict to cells of the hematopoietic system. It is expressed on monocytes, myeloid progenitors, granulocytes, mast cells, some T cells. This molecular may mediate cell-to-cell adhesion and act as receptor that inhibits the proliferation of normal and leukemic myeloid cells.

Western Blot:K-562 Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Specifications
  • Size:
    150 µL
  • Clonality:
    Polyclonal
  • Purification:
    Antigen affinity Purification
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Cat. No.:
    10084-332
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Shipping Temperature:
    Store at -20°C
  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD33
  • Immunogen:
    Recombinant Protein
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen Synonyms:
    SIGLEC-3|SIGLEC3|CD33|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3|p67
  • Antigen Name:
    CD33 molecule
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.