Anti-ENPP4 Rabbit Polyclonal Antibody
Catalog # 10086-540
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Ectonucleotide pyrophosphatase/phosphodiesterase 4
- Antigen Symbol:ENPP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:NPP4
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10086-540
- Supplier no.:17305-1-AP
Specifications
About this item
ENPP4, also named as KIAA0879 or NPP4, is a 453 amino acid protein, which belongs to the nucleotide pyrophosphatase/phosphodiesterase family. ENPP4 localizes in the cell membrane and hydrolyzes extracellular Ap3A into AMP and ADP, and Ap4A into AMP and ATP. ENPP4 is expressed on the surface of vascular endothelia.
Western Blot: HL-60 Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200
Type: Primary
Antigen: ENPP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat