Anti-NIP30 Rabbit Polyclonal Antibody
Catalog # 10091-108
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:NIP30
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10091-108
- Supplier no.:16830-1-AP
Specifications
About this item
NIP30, also known as NEFA-interacting nuclear protein 30, FAM192A (family with sequence similarity 192, member A), is a 254 amino acid nuclear protein encoded by a gene that maps to human chromosome 16q13.
Western Blot: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: NIP30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat