Order Entry
ContactUsLinkComponent
Anti-ZC3HAV1 Rabbit Polyclonal Antibody
Anti-ZC3HAV1 Rabbit Polyclonal Antibody
Catalog # 10097-254
Supplier:  Proteintech
Anti-ZC3HAV1 Rabbit Polyclonal Antibody
Catalog # 10097-254
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    ZC3HAV1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10097-254
  • Supplier no.:
    16820-1-AP

Specifications

About this item

The zinc-finger antiviral protein (ZAP) was originally identifed from a rat cDNA library due to its conferring resistance to retroviruses. ZC3HAV1 is a CCCH-type zinc finger protein, it may primarily function to inhibit viral gene expression and induce an innate immunity to viral infection. Alternative splicing occurs at this locus and two variants, each encoding distinct isoforms.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ZC3HAV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Frequently Bought Together