Order Entry
United States
ContactUsLinkComponent
Anti-EED Rabbit Polyclonal Antibody
Anti-EED Rabbit Polyclonal Antibody
Catalog # 10086-268
Supplier:  Proteintech
Anti-EED Rabbit Polyclonal Antibody
Catalog # 10086-268
Supplier:  Proteintech
Supplier Number:  16818-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Polycomb protein EED
  • Antigen Symbol:
    EED
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    HEED|WD protein associating with integrin cytoplasmic tails 1|WAIT1|polycomb protein EED
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10086-268
  • Supplier no.:
    16818-1-AP

Specifications

About this item

EED is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein interacts with enhancer of zeste 2, the cytoplasmic tail of integrin beta7, immunodeficiency virus type 1 (HIV-1) MA protein, and histone deacetylase proteins. This protein mediates repression of gene activity through histone deacetylation, and may act as a specific regulator of integrin function. EED has three isoforms, with the molecular weight of isoforms 1/2/3: 50/53/46 kDa.

Western Blot:COLO 320 Cells, 1:500-1:5000

Type: Primary
Antigen: EED
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat