Anti-CCKBR Rabbit Polyclonal Antibody
Catalog # 10084-214
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:CCKBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Peptide
- Purification:Antigen affinity Purification
- Cat. No.:10084-214
- Supplier no.:16549-1-AP
Specifications
About this item
Western Blot:BxPC-3 Cells, 1:200-1:2000; IF:BxPC-3 Cells, 1:10-1:100
Type: Primary
Antigen: CCKBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human