Anti-NUP160 Rabbit Polyclonal Antibody
Catalog # 10091-408
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Nucleoporin 160kDa
- Antigen Symbol:NUP160
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:160 kDa nucleoporin|NU160_HUMAN.|Nuclear pore complex protein Nup160|NUP120|Nucleoporin 160kDa|Nucleoporin 160kD|Nucleoporin Nup160|NUP 160
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10091-408
- Supplier no.:16084-1-AP
Specifications
About this item
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: NUP160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat