Anti-PCIF1 Rabbit Polyclonal Antibody
Catalog # 10091-442
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:PDX1 C-terminal inhibiting Factor 1
- Antigen Symbol:PCIF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10091-442
- Supplier no.:16082-1-AP
Specifications
About this item
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200
Type: Primary
Antigen: PCIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse