Anti-ARD1B Rabbit Polyclonal Antibody
Catalog # 10082-308
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:ARD1B
- Antigen Symbol:ARD1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-308
- Supplier no.:15670-1-AP
Specifications
About this item
ARD1B, also named as NAA11 and ARD2, belongs to the acetyltransferase family and ARD1 subfamily. In complex with NAA15, ARD1B displays alpha (N-terminal) acetyltransferase activity. It is tissue-specific and is epigenetically regulated by DNA methylation.
Western Blot: Mouse Testis Tissue, 1:500-1:5000
Type: Primary
Antigen: ARD1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat