Anti-URAT1 Rabbit Polyclonal Antibody
Catalog # 10096-594
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:URAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-594
- Supplier no.:14937-1-AP
Specifications
About this item
Human urate anion exchanger (URAT1), also known as SLC22A12, is a urate transporter located in the proximal tubules of human kidney and responsible for urate reabsorption. Defects in gene encoding URAT1 cause hypouricemia renal type 1 (RHUC1). This antibody recognizes the native and glycosylated URAT1 between 65 and 70 kDa as well as the truncated 40kDa protein.
Western Blot:PC-13 Cells, 1:200-1:2000
Type: Primary
Antigen: URAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat