Anti-AIFM3 Rabbit Polyclonal Antibody
10082-636
- Antibody Type:Primary
- Antigen Name:Apoptosis-inducing Factor, mitochondrion-associated, 3
- Antigen Symbol:AIFM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:AIFM3
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-636
- Supplier no.:14778-1-AP
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200
Type: Primary
Antigen: AIFM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat