Anti-SEC11A Rabbit Polyclonal Antibody
Catalog # 10094-242
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:SEC11 homolog A, signal peptidase complex subunit
- Antigen Symbol:SEC11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:SPCS4A|SEC11L1|1810012E07Rik|SPC18|sid2895
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10094-242
- Supplier no.:14753-1-AP
Specifications
About this item
SEC11A(Signal peptidase complex catalytic subunit SEC11A) is also named as SEC11L1, SPC18, SPCS4A and belongs to the peptidase S26B family. It is a component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum. The gene encode a 179 amino acid protein with 21 kDa.
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200
Type: Primary
Antigen: SEC11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat