- Antibody Type:Primary
- Antigen Name:Syntaxin 12
- Antigen Symbol:STX12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:MGC51957|STX12|STX14|STX13
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10095-520
- Supplier no.:14259-1-AP
Syntaxin 12 (STX12), also known as STX13 or STX14, is a 276 amino acid single-pass type IV membrane protein that belongs to the syntaxin family. It is a broadly and differentially expressed SNARE protein, highly enriched in brain tissue. STX12 is localized to early and recycling endosomes and is involved in the recycling of plasma membrane proteins. It also has a role in the trafficking of MMP during degradation of ECM substrates and subsequent cellular invasion. This antibody recognizes a major 39 kDa band of endogenous STX12. In some cases, two additional bands of 33 kDa and 66 kDa could also be detected, which most likely represent a degradation product and a SNARE complex respectively .
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A
Type: Primary
Antigen: STX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat